accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOHFFF

CAS RN 216766-12-0

IUPAC Name: 6-(trifluoromethyl)pyridin-3-ol
Synonyms: 6-(trifluoromethyl)pyridin-3-ol
Molecular Weight (g/mol): 163.1
Molecular Formula: C6H4F3NO
InChi Key: VFOBDHYPESAMAF-UHFFFAOYSA-N
SMILES: OC1=CN=C(C=C1)C(F)(F)F
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)