Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hydroxy-2-(trifluoromethyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. H14061G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H14061G Supplier TCI America Supplier No. H14061G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity organic building block with ≥98.0% purity.

Chemical Identifiers

CAS 216766-12-0
Molecular Formula C6H4F3NO
Molecular Weight (g/mol) 163.10
MDL Number MFCD06801325
InChI Key VFOBDHYPESAMAF-UHFFFAOYSA-N
Synonym 6-trifluoromethyl pyridin-3-ol, 5-hydroxy-2-trifluoromethyl pyridine, 6-trifluoromethyl-3-pyridinol, 2-trifluoromethyl pyridin-5-ol, 5-hydroxy-2-trifluoromethylpyridine, 3-pyridinol, 6-trifluoromethyl, 6-trifluoromethyl-pyridin-3-ol, pubchem21818, 6-trifluoromethyl 3-pyridinol
PubChem CID 2783388
IUPAC Name 6-(trifluoromethyl)pyridin-3-ol
SMILES OC1=CN=C(C=C1)C(F)(F)F

Specifications

Melting Point 174°C
Quantity 1 g
Formula Weight 163.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxy-2-(trifluoromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.