accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21679-31-2

IUPAC Name: chromium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Synonyms: chromium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular Weight (g/mol): 349.32
Molecular Formula: C15H21CrO6
InChi Key: JWORPXLMBPOPPU-LNTINUHCSA-K
SMILES: [Cr+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Quantity 
  • (5)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (7)
Physical Form 
  • (2)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (5)
  • (5)
  • (1)
  • (2)
  • (3)
  • (3)
  • (7)
  • (2)
  • (3)
  • (3)
  • (7)