Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromic Acetylacetonate, MP Biomedicals

Catalog No. ICN208809 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN208809 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN208809 Supplier MP Biomedicals, Inc Supplier No. 0520880980

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 21679-31-2
Molecular Formula C15H21CrO6
Molecular Weight (g/mol) 349.32
MDL Number MFCD00000015 MFCD00000015
InChI Key JWORPXLMBPOPPU-LNTINUHCSA-K
Synonym chromium iii acetylacetonate, dsstox_cid_30421, dsstox_gsid_51863
PubChem CID 91759531
IUPAC Name chromium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
SMILES [Cr+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Quantity 100 g
Formula Weight 349.32
Chemical Name or Material Chromic Acetylacetonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.