accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOOOHO

CAS RN 21691-43-0

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}pentanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}pentanoic acid|Z-DL-NVA-OH
Molecular Weight (g/mol): 251.28
Molecular Formula: C13H17NO4
InChi Key: NSJDRLWFFAWSFP-UHFFFAOYSA-N
SMILES: CCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)