Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-DL-norvaline 98.0+%, TCI America™
SDP

Catalog No. C07525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07525G 5 g
C075225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07525G Supplier TCI America Supplier No. C07525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 21691-43-0
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.28
MDL Number MFCD00020434
InChI Key NSJDRLWFFAWSFP-UHFFFAOYSA-N
Synonym z-dl-nva-oh, n-carbobenzoxy-dl-norvaline, n-cbz-dl-norvaline, n-benzyloxy carbonyl norvaline, n-cbz-l-norvaline, 2-benzyloxy carbonyl amino pentanoic acid, 2-phenylmethoxycarbonylamino pentanoic acid, dl-norvaline, n-phenylmethoxy carbonyl, z-dl-norvaline, cbz-dl-norvaline
PubChem CID 263473
IUPAC Name 2-{[(benzyloxy)carbonyl]amino}pentanoic acid
SMILES CCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 88°C
Color White
Quantity 5 g
Formula Weight 251.28
Percent Purity ≥98.0% (T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-DL-norvaline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.