CAS RN 216959-34-1
CAS RN 216959-34-1
IUPAC Name:
ethyl 2-{[(tert-butoxy)carbonyl]amino}-2-oxoacetate
Synonyms:
ethyl 2-[(tert-butoxycarbonyl)amino]-2-oxoacetate
Molecular Weight (g/mol):
217.22
Molecular Formula:
C9H15NO5
InChi Key:
DSDQWJVXMDHQLK-UHFFFAOYSA-N
SMILES:
CCOC(=O)C(=O)NC(=O)OC(C)(C)C
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich (+)-Trans-Chrysanthemic Acid
Sigma Aldrich (+)-Trans-Chrysanthemic Acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.