Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+)-Trans-Chrysanthemic Acid

Catalog No. 18509500MGF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-509-500MGF 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 18-509-500MGF Supplier Sigma Aldrich Supplier No. 18509500MGF
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(+)-trans-Chrysanthemic acid is the acidic component of synthetic pyrethroid insecticides. It can be prepared from (+)-+÷3-carene.(+)-trans-Chrysanthemic acid may be used in the preparation of (+)-trans-pyrethric acid, a building block for rethrin II.Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. 18509.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

Specifications

CAS 216959-34-1
Molecular Formula C9H15NO5
Refractive Index n20/D 1.477
Linear Formula C9H15NO5
MDL Number MFCD01631201
Quantity 5 g
Synonym (1R,3R)-trans-2,2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropane-1-carboxylic acid
Molecular Weight (g/mol) 217.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.