accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 216959-91-0

IUPAC Name: 4-(pyrimidin-5-yl)benzoate
Synonyms: 4-(pyrimidin-5-yl)benzoate
Molecular Weight (g/mol): 199.19
Molecular Formula: C11H7N2O2
InChi Key: CCBKADQVSXLSDN-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(C=C1)C1=CN=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results