accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3C

CAS RN 2170-08-3

IUPAC Name: dimethylbis(2-phenylethynyl)silane
Synonyms: dimethylbis(2-phenylethynyl)silane
Molecular Weight (g/mol): 260.41
Molecular Formula: C18H16Si
InChi Key: LRBLIVYQOCFXPX-UHFFFAOYSA-N
SMILES: C[Si](C)(C#CC1=CC=CC=C1)C#CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results