Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylbis(phenylethynyl)silane 98.0+%, TCI America™
SDP

Catalog No. D46255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46255G 5 g
D46251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46255G Supplier TCI America Supplier No. D46255G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2170-08-3
Molecular Formula C18H16Si
Molecular Weight (g/mol) 260.41
MDL Number MFCD00270967
InChI Key LRBLIVYQOCFXPX-UHFFFAOYSA-N
PubChem CID 11010735
IUPAC Name dimethyl-bis(2-phenylethynyl)silane
SMILES C[Si](C)(C#CC1=CC=CC=C1)C#CC2=CC=CC=C2

Specifications

Melting Point 81°C
Color Yellow
Boiling Point 177°C
Quantity 5 g
Formula Weight 260.41
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethylbis(phenylethynyl)silane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.