accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3HOCH3CH3CH3HOCH3CH3CH3HOCH3CH3CH3HOHf

CAS RN 2172-02-3

IUPAC Name: tetrakis(2-methylpropan-2-ol) hafnium
Synonyms: tetrakis(2-methyl-2-propanol) hafnium
Molecular Weight (g/mol): 474.98
Molecular Formula: C16H40HfO4
InChi Key: YZABPPRMRYHJGL-UHFFFAOYSA-N
SMILES: [Hf].CC(C)(C)O.CC(C)(C)O.CC(C)(C)O.CC(C)(C)O
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results