Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hafnium tert-butoxide, 99.9% (metals basis excluding Zr), Zr< 0.5%

Catalog No. p-7051750
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4190809 10 g
AA4190818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4190809 Supplier Thermo Scientific Chemicals Supplier No. 04190809
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2172-02-3 | C16H40HfO4 | 474.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2172-02-3
MDL Number MFCD00070458

Specifications

Melting Point 2°C
Color Yellow
Density 1.15 g/mL
Boiling Point 90°C (5 mmHg)
Flash Point 28°C (82°F)
Molecular Formula C16H40HfO4
Linear Formula Hf[OC(CH3)3]4
Quantity 10 g
UN Number UN1993
Sensitivity Air and moisture sensitive
Synonym tetrakis tert-butoxy hafnium, hafnium; 2-methylpropan-2-ol
Solubility Information Soluble in hydrocarbons, reacts with alcohols, ketones, and esters
InChI Key YZABPPRMRYHJGL-UHFFFAOYSA-N
SMILES CC(C)(C)O.CC(C)(C)O.CC(C)(C)O.CC(C)(C)O.[Hf]
IUPAC Name hafnium;2-methylpropan-2-ol
Molecular Weight (g/mol) 474.98
PubChem CID 25199745
Formula Weight 470.94
Percent Purity 99.9%
Physical Form Liquid
Assay (metals basis excluding Zr), Zr < 0.5%
Chemical Name or Material Hafnium tert-butoxide
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.