accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2173-57-1

IUPAC Name: 2-(2-methylpropoxy)naphthalene
Synonyms: isobutyl 2-naphthyl ether
Molecular Weight (g/mol): 200.28
Molecular Formula: C14H16O
InChi Key: XOHIHZHSDMWWMS-UHFFFAOYSA-N
SMILES: CC(C)COC1=CC=C2C=CC=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results