accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21778-19-8

IUPAC Name: (4-methoxyphenyl)phosphonic acid
Synonyms: 4-methoxyphenylphosphonic acid
Molecular Weight (g/mol): 188.12
Molecular Formula: C7H9O4P
InChi Key: SCMAYTUWDLAAAO-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)P(O)(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results