Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenylphosphonic Acid 95.0+%, TCI America™
SDP

Catalog No. M11551G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M11551G Supplier TCI America Supplier No. M11551G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 21778-19-8
Molecular Formula C7H9O4P
Molecular Weight (g/mol) 188.119
MDL Number MFCD00013948
InChI Key SCMAYTUWDLAAAO-UHFFFAOYSA-N
Synonym 4-Phosphonylanisole
PubChem CID 2779449
IUPAC Name (4-methoxyphenyl)phosphonic acid
SMILES COC1=CC=C(C=C1)P(=O)(O)O

Specifications

Quantity 1 g
Formula Weight 188.12
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenylphosphonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.