accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3C

CAS RN 21789-36-6

IUPAC Name: 2,4-dimethylbenzonitrile
Synonyms: 2,4-dimethylbenzonitrile
Molecular Weight (g/mol): 131.18
Molecular Formula: C9H9N
InChi Key: QLZDTHTXOUOSCV-UHFFFAOYSA-N
SMILES: CC1=CC=C(C#N)C(C)=C1
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)