Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethylbenzonitrile 98.0+%, TCI America™
SDP

Catalog No. D44955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44955G 5 g
D449525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44955G Supplier TCI America Supplier No. D44955G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 21789-36-6
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00016337
InChI Key QLZDTHTXOUOSCV-UHFFFAOYSA-N
Synonym 4-Cyano-m-xylene
PubChem CID 89046
IUPAC Name 2,4-dimethylbenzonitrile
SMILES CC1=CC(=C(C=C1)C#N)C

Specifications

Color White-Yellow
Quantity 5 g
Physical Form Crystal-Lump at 20°C
Boiling Point 109°C
Formula Weight 131.18
Percent Purity ≥98.0% (GC)
Grade Not Available
Chemical Name or Material 2,4-Dimethylbenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.