accessibility menu, dialog, popup

UserName

Viewing profile as user:
CaOHOHSOOOOHOHSOOO

CAS RN 21799-87-1

IUPAC Name: calcium bis(2,5-dihydroxybenzene-1-sulfonate)
Synonyms: calcium dobesilate
Molecular Weight (g/mol): 418.4
Molecular Formula: C12H10CaO10S2
InChi Key: QGNBTYAQAPLTMX-UHFFFAOYSA-L
SMILES: [Ca++].OC1=CC=C(O)C(=C1)S([O-])(=O)=O.OC1=CC=C(O)C(=C1)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)