Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroquinone sulfonic acid, potassium salt, Thermo Scientific™

Catalog No. AC120920500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC120920500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC120920500 Supplier Thermo Scientific Chemicals Supplier No. 120920500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21799-87-1
Molecular Formula C6H5KO5S
Molecular Weight (g/mol) 228.259
InChI Key VKDSBABHIXQFKH-UHFFFAOYSA-M
Synonym potassium 2,5-dihydroxybenzenesulfonate, hydroquinonesulfonic acid potassium salt, potassium2,5-dihydroxybenzenesulfonate, unii-249e3f00ep, benzenesulfonic acid, 2,5-dihydroxy-, monopotassium salt, 2,5-dihydroxybenzenesulfonic acid potassium salt, potassium hydroquinonesulfonate, hydroquinone sulfonic acid potassium salt, hydroquinone sulfonic acid, potassium salt, benzenesulfonic acid, 2,5-dihydroxy-, potassium salt 1:1
PubChem CID 23672329
IUPAC Name potassium;2,5-dihydroxybenzenesulfonate
SMILES C1=CC(=C(C=C1O)S(=O)(=O)[O-])O.[K+]

Specifications

Color Gray to White
Infrared Spectrum Authentic
Assay Percent Range 95%
Linear Formula (HO)2C6H3SO3K
Quantity 50 g
Beilstein 11, 300
Formula Weight 228.26
Percent Purity ≥94%
Physical Form Fine Crystalline Powder
Chemical Name or Material Hydroquinone sulfonic acid, potassium salt, tech., 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.