accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)N(R)(R)NCH3OCH3HO(R)OH3COOOCH3(R)

CAS RN 2182-14-1

IUPAC Name: methyl (1R,9R,11R,12R,19R)-11-(acetyloxy)-12-ethyl-10-hydroxy-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.0¹,⁹.0²,⁷.0¹⁶,¹⁹]nonadeca-2,4,6,13-tetraene-10-carboxylate
Synonyms: vindoline
Molecular Weight (g/mol): 456.54
Molecular Formula: C25H32N2O6
InChi Key: CXBGOBGJHGGWIE-MXXIQHGTSA-N
SMILES: CC[C@@]12C=CCN3CC[C@@]4([C@H]13)[C@@H](N(C)C1=CC(OC)=CC=C41)C(O)([C@@H]2OC(C)=O)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results