Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Vindoline

Catalog No. AC462270050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462270050 5 g
AC462270010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462270050 Supplier Thermo Scientific Chemicals Supplier No. 462270050
  • $554.85
    $471.65 / Each of 1

    Save $83.20

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2182-14-1 | C25H32N2O6 | 456.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2182-14-1
Molecular Formula C25H32N2O6
Molecular Weight (g/mol) 456.53
InChI Key CXBGOBGJHGGWIE-VJAGLATISA-N
Synonym vindoline
PubChem CID 129316872
SMILES CCC12C=CCN3C1C4(CC3)C(C(C2OC(=O)C)(C(=O)OC)O)N(C5=C4C=CC(=C5)OC)C

Specifications

Quantity 5 g
Formula Weight 456.53
Chemical Name or Material Vindoline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of causing genetic defects.
EINECSNumber 218-558-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.