accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3NHNOOOOH3CCH3

CAS RN 21829-25-4

IUPAC Name: 3,5-dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Synonyms: nifedipine anpine nificard ecodipin duranifin fenamon fedcor nifedine bonacid calcilat
Molecular Weight (g/mol): 346.34
Molecular Formula: C17H18N2O6
InChi Key: HYIMSNHJOBLJNT-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC=C1[N+]([O-])=O)C(=O)OC
Molecular Weight (g/mol) 
  • (5)
  • (8)
Percent Purity 
  • (2)
  • (5)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (6)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
Grade 
  • (1)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (5)
  • (1)
  • (1)
  • (2)
  • (6)
  • (1)
  • (1)
  • (5)
  • (8)
  • (2)
  • (5)
  • (3)
  • (3)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)