accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNH3OO

CAS RN 2185-03-7

IUPAC Name: 2-oxo-2,3-dihydrofuran-3-aminium chloride
Synonyms: 2-oxo-3H-furan-3-aminium chloride
Molecular Weight (g/mol): 135.55
Molecular Formula: C4H6ClNO2
InChi Key: SXCADCWKHNVANX-UHFFFAOYNA-N
SMILES: [Cl-].[NH3+]C1C=COC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)