Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-alpha-Amino-gamma-butyrolactone Hydrochloride 95.0+%, TCI America™
SDP

Catalog No. A14451G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A14451G Supplier TCI America Supplier No. A14451G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2185-03-7
Molecular Formula C4H6ClNO2
Molecular Weight (g/mol) 135.55
MDL Number MFCD00058172
InChI Key SXCADCWKHNVANX-UHFFFAOYNA-N
Synonym (S)-(-)-3-Aminotetrahydrofuran-2-one Hydrochloride, L-(-)-Homoserine Lactone Hydrochloride
PubChem CID 2733667
IUPAC Name (3S)-3-aminooxolan-2-one;hydrochloride
SMILES C1COC(=O)C1N.Cl

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 137.56
Percent Purity ≥95.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-alpha-Amino-gamma-butyrolactone Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.