accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21867-69-6

IUPAC Name: 4-[(4-methoxyphenyl)methyl]piperazine-1,4-diium
Synonyms: 4-[(4-methoxyphenyl)methyl]piperazine-1,4-diium
Molecular Weight (g/mol): 208.3
Molecular Formula: C12H20N2O
InChi Key: MGLUVVBFISROAH-UHFFFAOYSA-P
SMILES: COC1=CC=C(C[NH+]2CC[NH2+]CC2)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results