accessibility menu, dialog, popup

UserName

Viewing profile as user:
NCZnBr

CAS RN 218777-23-2

IUPAC Name: bromozincylium; pyridin-2-ide
Synonyms: bromozincylium; pyridin-2-ide
Molecular Weight (g/mol): 223.38
Molecular Formula: C5H4BrNZn
InChi Key: ZQJYTTPJYLKTTI-UHFFFAOYSA-M
SMILES: [Zn+]Br.C1=CC=N[C-]=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (1)
Quantity 
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)