Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pyridylzinc bromide, 0.5M solution in THF, AcroSeal™

Catalog No. AC433630500
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433630500 50 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC433630500 Supplier Thermo Scientific Chemicals Supplier No. 433630500
  • $341.00
    $274.65 / Each of 1

    Save $66.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 218777-23-2 | C5H4BrNZn | 223.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Pyridylzinc bromide
CAS 218777-23-2, 109-99-9
CAS Min % 10.32, 87.39
CAS Max % 12.61, 89.68
Color White
Density 0.9740g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C5H4BrNZn
Quantity 50 mL
Specific Gravity 0.974
Synonym 2-pyridylzinc bromide
InChI Key WUTSMSLAOBVVCP-UHFFFAOYSA-M
SMILES C1=CC=NC=C1.[Zn].[Br-]
IUPAC Name pyridine;zinc;bromide
Molecular Weight (g/mol) 223.39
PubChem CID 76056926
Formula Weight 223.39
Physical Form Crystalline Powder
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.