accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21898-19-1

IUPAC Name: hydrogen 1-(4-amino-3,5-dichlorophenyl)-2-(tert-butylamino)ethan-1-ol chloride
Synonyms: hydrogen clenbuterol chloride
Molecular Weight (g/mol): 313.65
Molecular Formula: C12H19Cl3N2O
InChi Key: OPXKTCUYRHXSBK-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(C)(C)NCC(O)C1=CC(Cl)=C(N)C(Cl)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results