Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-alpha-(tert-butylaminomethyl)-3,5-dichlorobenzyl Alcohol Hydrochloride 97.0+%, TCI America™
SDP

Catalog No. C2691100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2691100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2691100MG Supplier TCI America Supplier No. C2691100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 21898-19-1
Molecular Formula C12H19Cl3N2O
Molecular Weight (g/mol) 313.647
MDL Number MFCD00083280
InChI Key OPXKTCUYRHXSBK-UHFFFAOYSA-N
Synonym clenbuterol hydrochloride, clenbuterol hcl, spiropent, clenbuterol clorhidrato, ventipulmin, 1-4-amino-3,5-dichlorophenyl-2-tert-butylamino ethanol hydrochloride, 4-amino-alpha-t-butylaminomethyl-3,5-dichlorobenzyl alcohol hydrochloride, planipart hydrochloride, 4-amino-alpha-tert-butylaminomethyl-3,5-dichlorobenzyl alcohol hydrochloride
PubChem CID 5702273
ChEBI CHEBI:31410
IUPAC Name 1-(4-amino-3,5-dichlorophenyl)-2-(tert-butylamino)ethanol;hydrochloride
SMILES CC(C)(C)NCC(C1=CC(=C(C(=C1)Cl)N)Cl)O.Cl

Specifications

Melting Point 164°C
Color Yellow
Quantity 100 mg
UN Number 2811
Formula Weight 313.65
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-alpha-(tert-butylaminomethyl)-3,5-dichlorobenzyl Alcohol Hydrochloride
RTECSNumber DN3180000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.