accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21934-68-9

IUPAC Name: 3'-(diethylamino)-6',8'-dimethyl-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms: 3'-(diethylamino)-6',8'-dimethylspiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular Weight (g/mol): 399.49
Molecular Formula: C26H25NO3
InChi Key: XUFBVJQHCCCPNM-UHFFFAOYNA-N
SMILES: CCN(CC)C1=CC=C2C(OC3=CC(C)=CC(C)=C3C22OC(=O)C3=CC=CC=C23)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results