Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6'-(Diethylamino)-1',3'-dimethylfluoran 98.0+%, TCI America™
SDP

Catalog No. D320225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D320225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D320225G Supplier TCI America Supplier No. D320225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 21934-68-9
Molecular Formula C26H25NO3
Molecular Weight (g/mol) 399.49
MDL Number MFCD08276349
InChI Key XUFBVJQHCCCPNM-UHFFFAOYNA-N
PubChem CID 89556
IUPAC Name 3'-(diethylamino)-6',8'-dimethyl-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
SMILES CCN(CC)C1=CC=C2C(OC3=CC(C)=CC(C)=C3C22OC(=O)C3=CC=CC=C23)=C1

Specifications

Melting Point 170°C
Color White-Yellow
Quantity 25 g
Formula Weight 399.49
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6′-(Diethylamino)-1′,3′-dimethylfluoran
EINECSNumber (5)-3618
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.