accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21947-32-0

OHOOOHN(S)H3CCH3CH3H3CHN

CAS RN 21947-32-0

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid; N-cyclohexylcyclohexanamine
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]hexanoic acid; dicha
Molecular Weight (g/mol): 412.62
Molecular Formula: C23H44N2O4
InChi Key: BFEVJIUEBDZIJQ-WDBKTSHHSA-N
SMILES: C1CCC(CC1)NC1CCCCC1.CCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)