accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21947-97-7

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-[(diphenylmethyl)sulfanyl]propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-[(diphenylmethyl)sulfanyl]propanoic acid
Molecular Weight (g/mol): 387.49
Molecular Formula: C21H25NO4S
InChi Key: AMTHPOXIVDHMLZ-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CSC(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)