accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21948-70-9

IUPAC Name: 2-(methylsulfanyl)pyrazine
Synonyms: 2-methylthiopyrazine
Molecular Weight (g/mol): 126.18
Molecular Formula: C5H6N2S
InChi Key: KBPBOWBQRUXMFV-UHFFFAOYSA-N
SMILES: CSC1=CN=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results