Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Methylthio)pyrazine 98.0+%, TCI America™
SDP

Catalog No. M182125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M182125G 25 g
M18215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M182125G Supplier TCI America Supplier No. M182125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 21948-70-9
Molecular Formula C5H6N2S
Molecular Weight (g/mol) 126.177
MDL Number MFCD00041024
InChI Key KBPBOWBQRUXMFV-UHFFFAOYSA-N
Synonym 2-methylthio pyrazine, 2-methylthiopyrazine, pyrazine, methylthio, pyrazinyl methyl sulfide, 2-methylsulfanyl pyrazine, methyl pyrazinyl sulfide, methylthio pyrazine, 2-methylsulfanyl-pyrazine, pyrazine, 2-methylthio, unii-r1c97e3a4f
PubChem CID 519907
IUPAC Name 2-methylsulfanylpyrazine
SMILES CSC1=NC=CN=C1

Specifications

Melting Point 45°C
Color White-Yellow
Quantity 25 g
Formula Weight 126.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Methylthio)pyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.