accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNHNFFF

CAS RN 219508-27-7

IUPAC Name: 2-[5-(trifluoromethyl)-1H-1,2,4-triazol-3-yl]pyridine
Synonyms: 2-[5-(trifluoromethyl)-1H-1,2,4-triazol-3-yl]pyridine
Molecular Weight (g/mol): 214.15
Molecular Formula: C8H5F3N4
InChi Key: MOGTVLAYAVGOJK-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=NC(=NN1)C1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results