Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[5-(Trifluoromethyl)-1H-1,2,4-triazol-3-yl]pyridine 98.0+%, TCI America™
SDP

Catalog No. T3638200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3638200MG 200 mg
T36381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3638200MG Supplier TCI America Supplier No. T3638200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 219508-27-7
Molecular Formula C8H5F3N4
Molecular Weight (g/mol) 214.151
MDL Number MFCD26403657
InChI Key MOGTVLAYAVGOJK-UHFFFAOYSA-N
Synonym 3-(Trifluoromethyl)-5-(2-pyridyl)-1,2,4-triazole
PubChem CID 20807129
IUPAC Name 2-[5-(trifluoromethyl)-1H-1,2,4-triazol-3-yl]pyridine
SMILES C1=CC=NC(=C1)C2=NNC(=N2)C(F)(F)F

Specifications

Melting Point 159°C
Color White-Yellow
Quantity 200 mg
Formula Weight 214.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-[5-(Trifluoromethyl)-1H-1,2,4-triazol-3-yl]pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.