accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCl

CAS RN 2196-99-8

IUPAC Name: 2-chloro-1-(4-methoxyphenyl)ethan-1-one
Synonyms: 2-chloro-1-(4-methoxyphenyl)ethanone
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: MCRINSAETDOKDE-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results