Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenacyl chloride, 97%

Catalog No. AC425260010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC425260010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC425260010 Supplier Thermo Scientific Chemicals Supplier No. 425260010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2196-99-8 | C9H9ClO2 | 184.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2196-99-8
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.62
MDL Number MFCD00216508
InChI Key MCRINSAETDOKDE-UHFFFAOYSA-N
Synonym 2-chloro-1-4-methoxyphenyl ethanone, 4-methoxyphenacyl chloride, 2-chloro-4'-methoxyacetophenone, 2-chloro-4'-methoxylphacetone, 4-methoxyphenacylchloride, 2-chloro-1-4-methoxy-phenyl-ethanone, 2-chloro-1-4-methoxyphenyl ethan-1-one, ethanone, 2-chloro-1-4-methoxyphenyl, 4-chloroacetyl anisole, 4'-methoxy-2-chloroacetophenone
PubChem CID 237806
IUPAC Name 2-chloro-1-(4-methoxyphenyl)ethanone
SMILES COC1=CC=C(C=C1)C(=O)CCl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 98°C to 100°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H3COC6H4COCH2Cl
Quantity 1 g
Solubility Information Solubility in water: negligible
Formula Weight 184.62
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenacyl chloride, 96.5%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.