accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONClI

CAS RN 219727-21-6

IUPAC Name: ethyl 2-chloro-4-iodopyridine-3-carboxylate
Synonyms: ethyl 2-chloro-4-iodopyridine-3-carboxylate
Molecular Weight (g/mol): 311.5
Molecular Formula: C8H7ClINO2
InChi Key: YOJVEUAQIMBZSL-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(I)C=CN=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results