Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-chloro-4-iodonicotinate, 97%, Thermo Scientific™

Catalog No. AAH5008503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5008503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5008503 Supplier Thermo Scientific Chemicals Supplier No. H5008503

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 219727-21-6
Molecular Formula C8H7ClINO2
Molecular Weight (g/mol) 311.50
MDL Number MFCD07781133
InChI Key YOJVEUAQIMBZSL-UHFFFAOYSA-N
Synonym ethyl 2-chloro-4-iodonicotinate, 3-pyridinecarboxylic acid, 2-chloro-4-iodo-, ethyl ester, 2-chloro-4-iodonicotinic acid ethyl ester, 2-chloro-4-iodo-nicotinic acid ethyl ester, 2-chloro-4-iodopyridine-3-carboxylic acid ethyl ester, 3-pyridinecarboxylicacid,2-chloro-4-iodo-,ethylester
PubChem CID 15850399
IUPAC Name ethyl 2-chloro-4-iodopyridine-3-carboxylate
SMILES CCOC(=O)C1=C(I)C=CN=C1Cl

Specifications

Quantity 1 g
Sensitivity Light sensitive
Formula Weight 311.51
Percent Purity 97%
Chemical Name or Material Ethyl 2-chloro-4-iodonicotinate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.