accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2198-59-6

IUPAC Name: hydrogen N1-phenylbenzene-1,4-diamine chloride
Synonyms: hydrogen N-phenyl-p-phenylenediamine chloride
Molecular Weight (g/mol): 220.7
Molecular Formula: C12H13ClN2
InChi Key: PNEVDCGZQWFIKV-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=CC=C(NC2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)