Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminodiphenylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. P018925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P018925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P018925G Supplier TCI America Supplier No. P018925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2198-59-6
Molecular Formula C12H13ClN2
Molecular Weight (g/mol) 220.7
MDL Number MFCD00012988
InChI Key PNEVDCGZQWFIKV-UHFFFAOYSA-N
Synonym n1-phenylbenzene-1,4-diamine hydrochloride, 4-aminodiphenylamine hydrochloride, eucanine grey b, 1,4-benzenediamine, n-phenyl-, monohydrochloride, fourrine grey b, durafur grey bn, fouramine grey a, futramine grey b, furro grey b, nako grey gb
PubChem CID 75146
IUPAC Name 4-N-phenylbenzene-1,4-diamine;hydrochloride
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)N.Cl

Specifications

Melting Point 241°C
Quantity 25 g
Formula Weight 220.70
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminodiphenylamine Hydrochloride
RTECSNumber ST3551000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.