accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHNONNNOOH3COOCH3OOCH3SOOH3C

CAS RN 219814-65-0

IUPAC Name: methyl 4-(acetyloxy)-6-[1,2-bis(acetyloxy)-3-azidopropyl]-5-acetamido-2-(phenylsulfanyl)oxane-2-carboxylate
Synonyms: methyl 4-(acetyloxy)-6-[1,2-bis(acetyloxy)-3-azidopropyl]-5-acetamido-2-(phenylsulfanyl)oxane-2-carboxylate
Molecular Weight (g/mol): 566.58
Molecular Formula: C24H30N4O10S
InChi Key: YCUSQTURLGBYSS-UHFFFAOYNA-N
SMILES: COC(=O)C1(CC(OC(C)=O)C(NC(C)=O)C(O1)C(OC(C)=O)C(CN=[N+]=[N-])OC(C)=O)SC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results