Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neu5Ac[1Me,478Ac,9N3]-β-SPh 92.0+%, TCI America™
SDP

Catalog No. M2695100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2695100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M2695100MG Supplier TCI America Supplier No. M2695100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 219814-65-0
Molecular Formula C24H30N4O10S
Molecular Weight (g/mol) 566.582
InChI Key YCUSQTURLGBYSS-DSHZTGCJSA-N
Synonym Methyl 5-Acetamido-4,7,8-tri-O-acetyl-9-azido-3,5,9-trideoxy-2-S-phenyl-2-thio-D-glycero-beta-D-galacto-2-nonulopyranosylonate, Neu5Ac[1Me,478Ac,9N3]-beta-SPh
PubChem CID 133556286
IUPAC Name methyl 5-acetamido-4-acetyloxy-6-[(1S,2S)-1,2-diacetyloxy-3-azidopropyl]-2-phenylsulfanyloxane-2-carboxylate
SMILES CC(=O)NC1C(CC(OC1C(C(CN=[N+]=[N-])OC(=O)C)OC(=O)C)(C(=O)OC)SC2=CC=CC=C2)OC(=O)C

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 566.58
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl (Phenyl 5-Acetamido-4,7,8-tri-O-acetyl-9-azido-3,5,9-trideoxy-2-thio-D-glycero-beta-D-galacto
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.