accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COFFFFFFOCH3FF(?)

CAS RN 2200-71-7

IUPAC Name: 2,2',3,3',5,5',6,6'-octafluoro-4,4'-dimethoxy-1,1'-biphenyl
Synonyms: 2,2',3,3',5,5',6,6'-octafluoro-4,4'-dimethoxy-1,1'-biphenyl
Molecular Weight (g/mol): 358.19
Molecular Formula: C14H6F8O2
InChi Key: TVUPJKALGKCABC-UHFFFAOYSA-N
SMILES: COC1=C(F)C(F)=C(C(F)=C1F)C1=C(F)C(F)=C(OC)C(F)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results