Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dimethoxyoctafluorobiphenyl 98.0+%, TCI America™
SDP

Catalog No. D23305G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23305G 5 g
D23301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23305G Supplier TCI America Supplier No. D23305G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2200-71-7
Molecular Formula C14H6F8O2
Molecular Weight (g/mol) 358.19
MDL Number MFCD00039802
InChI Key TVUPJKALGKCABC-UHFFFAOYSA-N
PubChem CID 262954
IUPAC Name 2,2',3,3',5,5',6,6'-octafluoro-4,4'-dimethoxy-1,1'-biphenyl
SMILES COC1=C(F)C(F)=C(C(F)=C1F)C1=C(F)C(F)=C(OC)C(F)=C1F

Specifications

Melting Point 87°C
Color White-Yellow
Quantity 5 g
Synonym Octafluoro-4,4′-dimethoxybiphenyl
Formula Weight 358.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dimethoxyoctafluorobiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.