CAS RN 220036-08-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ ATR/CDK Inhibitor, NU6027
The ATR/CDK Inhibitor, NU6027, also referenced under CAS 220036-08-8, controls the biological activity of ATR/CDK. This small molecule/inhibitor is primarily used for Cancer applications.