accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 220141-74-2

IUPAC Name: 2-(2,4,5-trifluorophenyl)acetonitrile
Synonyms: 2-(2,4,5-trifluorophenyl)acetonitrile
Molecular Weight (g/mol): 171.12
Molecular Formula: C8H4F3N
InChi Key: JTYBTJVFXUKNKW-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(CC#N)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results